Få mere at vide
Beta-Chloro-DL-alanine-d3, TRC
Artikelnummer. 30360047
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30360047 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360047 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3641922.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Chloro-DL-alanine-d3
CAS 2124271-38-9
Molekylær formel C3H3D3ClNO2
Mængde 2.5 mg
Synonym 3-Chloro-DL-alanine-d3, 3-Chloroalanine-d3, DL-3-Chloroalanine-d3, β-Chloro-DL-alanine-d3, β-Chloroalanine-d3
InChI formel InChI=1S/C3H6ClNO2/c4-1-2(5)3(6)7/h2H,1,5H2,(H,6,7)/i1D2,2D
SMIL ClC([2H])([2H])C(N)([2H])C(O)=O
IUPAC navn 2-amino-3-chloropropanoic-2,3,3-d3 acid
Molekylvægt (g/mol) 126.55
Formel vægt 126.03