Få mere at vide
(-)-Beta-Bisabolene (>85%), TRC
Artikelnummer. 30135816
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135816 50 mg
30010522 25 mg
30009203 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135816 Leverandør Toronto Research Chemicals Leverandørnr. TRCB39980550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 495-61-4
Molekylær formel C15 H24
Molekylvægt (g/mol) 204.35
Synonym Cyclohexene, 1-methyl-4-(5-methyl-1-methylene-4-hexen-1-yl)-, (4S)-, 1,5-Heptadiene, 6-methyl-2-(4-methyl-3-cyclohexen-1-yl)-, (S)-(-)- (8CI), Cyclohexene, 1-methyl-4-(5-methyl-1-methylene-4-hexenyl)-, (4S)- (9CI), Cyclohexene, 1-methyl-4-(5-methyl-1-methylene-4-hexenyl)-, (S)-, β-Bisabolene (6CI), (4S)-1-Methyl-4-(5-methyl-1-methylene-4-hexen-1-yl)cyclohexene, (-)-β-Bisabolene, (S)-1-Methyl-4-(5-methyl-1-methylene-4-hexenyl)-1-cyclohexene, (S)-1-Methyl-4-(5-methyl-1-methylene-4-hexenyl)cyclohexene, (S)-β-Bisabolene, l-β-Bisabolene
IUPAC navn (4S)-1-methyl-4-(6-methylhepta-1,5-dien-2-yl)cyclohexene
SMIL CC(=CCCC(=C)[C@H]1CCC(=CC1)C)C
Tekniske data
Kemisk navn eller materiale (-)-beta-Bisabolene
Mængde 50 mg
InChI formel InChI=1S/C15H24/c1-12(2)6-5-7-14(4)15-10-8-13(3)9-11-15/h6,8,15H,4-5,7,9-11H2,1-3H3/t15-/m1/s1
Formel vægt 204.1878