Få mere at vide
Beta-alanin ethylesterhydrochlorid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | beta-Alanine Ethyl Ester Hydrochloride |
| CAS | 4244-84-2 |
| Molekylær formel | C5H12ClNO2 |
| Mængde | 5 g |
| Synonym | β-Alanine ethyl ester hydrochloride (1:1), β-Alanine, ethyl ester, hydrochloride (6CI,7CI,8CI,9CI), 3-Aminopropanoic acid ethyl ester hydrochloride, 3-Aminopropionic acid ethyl ester hydrochloride, CarnoSyn, Ethyl 3-aminopropanoate hydrochloride, Ethyl 3-aminopropionate hydrochloride, Ethyl β-alaninate hydrochloride, Ethyl β-alaninoate hydrochloride |
| InChI formel | InChI=1S/C5H11NO2.ClH/c1-2-8-5(7)3-4-6;/h2-4,6H2,1H3;1H |
| SMIL | Cl.CCOC(=O)CCN |
| IUPAC navn | ethyl3-aminopropanoat; hydrochlorid |
| Molekylvægt (g/mol) | 153.61 |
| Formel vægt | 153.0557 |