Learn More
beta-Alanine ethyl ester hydrochloride, 98%
CAS: 4244-84-2 | C5H12ClNO2 | 153.61 g/mol
Brand: Thermo Scientific Alfa Aesar A17277.18
1305.67 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Quantity | 50 g |
---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4244-84-2 | |
153.61 | |
RJCGNNHKSNIUAT-UHFFFAOYSA-N | |
458475 | |
Cl.CCOC(=O)CCN |
C5H12ClNO2 | |
MFCD00012909 | |
ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl | |
ethyl 3-aminopropanoate;hydrochloride |
Specifications
67°C to 72°C | |
C5H12ClNO2 | |
3559095 | |
RJCGNNHKSNIUAT-UHFFFAOYSA-N | |
ethyl 3-aminopropanoate;hydrochloride | |
50 g | |
153.61 | |
98% |
4244-84-2 | |
MFCD00012909 | |
ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl | |
Cl.CCOC(=O)CCN | |
153.61 | |
458475 | |
Hygroscopic | |
beta-Alanine ethyl ester hydrochloride |