missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alaninamide hydrochloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat , air and oxidizing agents.
beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat , air and oxidizing agents.
Specifications
Melting Point | 149°C to 154°C |
Beilstein | 3909917 |
Synonym | 3-aminopropanamide hydrochloride, 3-aminopropionamide hydrochloride, 3-aminopropanamide hcl, beta-alaninamide hydrochloride, h-beta-ala-nh2.hcl, h-beta-ala-nh2 hcl, 3-amino-propionamide hydrochloride, h-ala-nh2hcl, propanamide, 3-amino-, monohydrochloride, h-beta-ala-nh2hcl |
Solubility Information | Soluble in methanol. |
Formula Weight | 124.57 |
Percent Purity | 96% |
Quantity | 5 g |
Sensitivity | Air sensitive |
Physical Form | Solid |
Chemical Name or Material | beta-Alaninamide hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'