Få mere at vide
beta Actin Rabbit anti-Human, Mouse, Non-human primate, Rat, Zebrafish, Polyclonal, Proteintech
Artikelnummer. 16840085 Shop alle Proteintech produkter
Skift visning
Mængde:
150 μL
Artikelnummer. Mængde
16840085 150 μL
1 options
Artikelnummer. 16840085 Leverandør Proteintech Leverandørnr. 205361AP150UL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
This antibody was generated against N-terminal region of human beta actin protein and can cross-react with other actins.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
Tekniske data
Antigen beta Actin
Applikationer Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Koncentration 0.43 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACTA1
Genadgangsnr. P60709, P60710, P60711, P62736, P62737, P62738, P63259, P63260, P63261, P63267, P63268, P63269, P68032, P68033, P68035, P68133, P68134, P68136
Gene Alias ACTB, Actin, actin, beta, Actin, cytoplasmic 1, B actin, Beta actin, F actin, PS1TP5BP1, beta actin
Gensymboler Acta1, acta1b, acta2, ACTB, actb1, Actc1, actc1b, Actg1, Actg2
Værtsarter Rabbit
Immunogen beta Actin Fusion Protein Ag14521
Oprensningsmetode Antigen Affinity Chromatography
Mængde 150 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 11459, 11461, 11464, 11465, 11468, 11475, 25365, 287876, 29275, 29437, 322509, 407658, 57934, 58, 58114, 59, 60, 70, 71, 72, 81633, 81822
Målarter Human, Mouse, Non-human Primate, Rat, Zebrafish
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG