Få mere at vide
Bestatin hydrochloride, MedChemExpress
Artikelnummer. 16494247
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16494247 50 mg
16478358 10 mg
16448126 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16494247 Leverandør Medchem Express Leverandørnr. HYB0134A
MedChemExpress Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer research.
Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer research.
Tekniske data
Kemisk navn eller materiale Bestatin hydrochloride
Fysisk form Solid
CAS 65391-42-6
Farve White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C16H25ClN2O4
Til brug med (applikation) COVID-19-immunoregulation
Synonym Ubenimex hydrochloride
Opløselighedsinformation DMSO : 250 mg/mL (724.99 mM; Need ultrasonic) ∣H2O : 100 mg/mL (290.00 mM; Need ultrasonic)
SMIL CC(C)C[C@@H](C(O)=O)NC([C@@H](O)[C@H](N)CC1=CC=CC=C1)=O.Cl
Molekylvægt (g/mol) 344.83
Formel vægt 344.83
Procent renhed 99.17%