Learn More
SHOWA™ 874 Best™ Butyl II Gloves
Same chemical permeation resistance
Brand: SHOWA™ 8635713
Additional Details : Weight : 0.80000kg
Description
The gloves are made with thinner construction for better dexterity.
- Available in both smooth and rough finish
Specifications
Black | |
Butyl | |
0.35 mm | |
Rolled | |
Chemicals including aldehydes, amines, and ketones |
Rough | |
10 | |
Chemicals handling | |
Unlined | |
355 mm |