Få mere at vide
MedChemExpress Beryllon II
Artikelnummer. 16481169
Encompass_Preferred
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
16481169 1 g
1 options
Artikelnummer. 16481169 Leverandør MedChemExpress Leverandørnr. HY112276
MedChemExpress Beryllon II is a widely used chromogenic reagent that is used to determine many elements, such as Mo, Mg and Co, and also used for the determination of proteins.
Tekniske data
CAS 51550-25-5
Molekylvægt (g/mol) 738.52 g/mol
Mængde 1 g
Forsendelsestilstand Room temperature
Opbevaringskrav 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Opløselighed DMSO : ≥ 64 mg/mL (86.66 mM)