Learn More
Berkshire™ 360 Isolator Cleaning System
Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Brand: Berkshire™
Additional Details : Weight : 2.00000kg
Description
EasyClean 360 ICT Sterile Knitted Polyester Elastic and Padded Covers (17-987-362)
- Made from Berkshire’s SuperPolx™ 1200A 100% continuous filament knitted polyester material
- Laundered and packaged in an ISO Class 4 cleanroom for superior giver, particle and residue performance
- Non-shedding and low-linting cover per USP requirements
- Stretch fabric and elastic edge for easy on/off with secure fit on mop head
- Absorbent polyester knit is abrasion resistant; handles a wide range of cleaning solutions
- Compatible for use in ISO Class 3/EU Grade A Environments
- Sterile
Specifications
Knitted Polyester Elastic Covers with Padded Cover, Sterile | |
EasyClean™ 360 Isolator Cleaning Tool | |
Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments. | |
Knitted polyester elastic |
10 sterile knitted polyester elastic covers and 1 sterile knitted polyester padded cover/pack; 20 packs/case | |
Mop Covers | |
ISO Class 3/EU Grade A |