missing translation for 'onlineSavingsMsg'
Få mere at vide

Bergamottin, TRC

Artikelnummer. 30347922
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347922 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30347922 Leverandør Toronto Research Chemicals Leverandørnr. TRCB318400100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Bergamottin
CAS 7380-40-7
Molekylær formel C21 H22 O4
Mængde 100 mg
Synonym 4-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-7H-Furo[3,2-g][1]benzopyran-7-one, 7H-Furo[3,2-g][1]benzopyran-7-one, 4-[(3,7-dimethyl-2,6-octadienyl)oxy]-, (E)- (8CI), 7H-Furo[3,2-g][1]benzopyran-7-one, 4-[[(2E)-3,7-dimethyl-2,6-octadienyl]oxy]- (9CI), Bergamottin (6CI), 4-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-7H-furo[3,2-g][1]benzopyran-7-one, 5-Geranoxypsoralen, Bergamotine, Bergaptin
InChI formel InChI=1S/C21H22O4/c1-14(2)5-4-6-15(3)9-11-24-21-16-7-8-20(22)25-19(16)13-18-17(21)10-12-23-18/h5,7-10,12-13H,4,6,11H2,1-3H3/b15-9+
SMIL CC(=CCC\C(=C\COc1c2C=CC(=O)Oc2cc3occc13)\C)C
IUPAC navn 4-[(2E)-3,7-dimethylocta-2,6-dienoxy]furo[3,2-g]chromen-7-one
Molekylvægt (g/mol) 338.4
Formel vægt 338.1518
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.