Learn More
Emma Safety Footwear Bergamo Safety Shoes
Comfortable foot-hugging fit provides a naturally light and airy foot feel. Virtually penetration proof, water resistant uppers. The 100% nitrile rubber outsole repels liquids, harmful or otherwise and the ingeniously designed sole contours provide extra grip on almost all surfaces.
POA
Specifications
Certifications/Compliance | EN ISO 20347:2012 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Bergamo |
Liner Material | Hydro-Tec™ Sanitized Silver & Wellness leather |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15998161
|
Emma Safety Footwear
107092-39 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908171
|
Emma Safety Footwear
107092-40 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15918171
|
Emma Safety Footwear
107092-41 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15928171
|
Emma Safety Footwear
107092-42 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15938171
|
Emma Safety Footwear
107092-44 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15948171
|
Emma Safety Footwear
107092-45 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15958171
|
Emma Safety Footwear
107092-46 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15968171
|
Emma Safety Footwear
107092-47 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978171
|
Emma Safety Footwear
107092-48 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- O3-Work shoe without steel toecap with sole plate
- Full grain leather construction
- Nitrile Rubber Heat Resistant Outsole
- Metal-free
- Chrome-free lining
- ESD certified
- Sanitized
Specifications
EN ISO 20347:2012 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather |
Laces | |
Bergamo | |
Leather |