missing translation for 'onlineSavingsMsg'
Få mere at vide

Berberine chloride hydrate, 96%, water <17%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar L03807.14

Mængde 25g

 Se flere versioner af dette produkt

Produktkode 11306666

  • 635.83 DKK / 25g

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

Solubility
Soluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY
Tekniske data

Kemiske identifikatorer

141433-60-5
389.832
BPNJXFPOPCFZOC-UHFFFAOYSA-M
155074
C20H20ClNO5
MFCD00149998
berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.[Cl-]

Tekniske data

Berberine chloride hydrate
water <17%
MFCD00149998
3836585
14,1154
Soluble in water.
COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.[Cl-]
155074
97%
141433-60-5
C20H20ClNO5
25g
Light Sensitive
berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
BPNJXFPOPCFZOC-UHFFFAOYSA-M
389.832
371.82 (Anhydrous)
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.