Få mere at vide
Tocris Bioscience™ Beraprost sodium
Beskrivelse
Beraprost sodium is a potent IP receptor agonist; analog of Epoprostenol (Cat. No. 2989). Potently inhibits ADP-induced platelet aggregation and P-selectin expression in vitro (pIC50 values are 8.26 and 8.56, respectively). Also increases vasodilation and reduces pulmonary hypertension in vivo. Orally active.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | rel-(1R,2R,3aS,8bS)-2,3,3a,8b-Tetrahydro-2-hydroxy-1-[(1E,3S)-3-hydroxy-4-methyl-1-octen-6-yn-1-yl)-1H-cyclopenta[b]benzofuran-5-butanoic acid sodium salt |
| CAS | 496807-11-5 |
| Mængde | 1 mg |
| Mål | Prostanoid Receptor Agonists |
| Molekylær formel | C24H29NaO5 |
| Formel vægt | 420.47 |
| Renhed | >98% |