Få mere at vide
Bephenium Hydroxynaphthoate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bephenium Hydroxynaphthoate |
| CAS | 3818-50-6 |
| Analyt- eller komponentnavne | Bephenium Hydroxynaphthoate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H22 N O . C11 H7 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzenemethanaminium, N,N-dimethyl-N-(2-phenoxyethyl)-, 3-hydroxy-2-naphthalenecarboxylate (1:1), Ammonium, benzyldimethyl(2-phenoxyethyl)-, salt with 3-hydroxy-2-naphthoic acid (1:1) (8CI), Benzenemethanaminium, N,N-dimethyl-N-(2-phenoxyethyl)-, salt with 3-hydroxy-2-naphthalenecarboxylic acid (1:1) (9CI), Benzyldimethyl(2-phenoxyethyl)ammonium 3-hydroxy-2-naphthoate (6CI,7CI), 2-Naphthalenecarboxylic acid, 3-hydroxy-, ion(1-), N,N-dimethyl-N-(2-phenoxyethyl)benzenemethanaminium (9CI), 2-Naphthoic acid, 3-hydroxy-, ion(1-), benzyldimethyl(2-phenoxyethyl)ammonium (8CI), Alcopar, Alcopara, Befen, Befeniol, Befenium, Bephenamine hydroxynaphthoate, Bephenium 3-hydroxy-2-naphthoate, Bephenium hydroxynaphthoate, Debefenium, Fedal-Uncin, Frantin, Lecibis, Naftamon, Naphtamon, Naphtamone, Nemex |
| InChI formel | InChI=1S/C17H22NO.C11H8O3/c1-18(2,15-16-9-5-3-6-10-16)13-14-19-17-11-7-4-8-12-17;12-10-6-8-4-2-1-3-7(8)5-9(10)11(13)14/h3-12H,13-15H2,1-2H3;1-6,12H,(H,13,14)/q+1;/p-1 |
| Løsningstype | Neat |
| SMIL | C[N+](C)(CCOc1ccccc1)Cc2ccccc2.Oc3cc4ccccc4cc3C(=O)[O-] |
| Vis mere |