Få mere at vide
Bephenium Embonate, LoGiCal
Artikelnummer. 30464001
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464001 10 mg
1 options
Artikelnummer. 30464001 Leverandør LoGiCal Leverandørnr. LGCFOR1833.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bephenium Embonate
CAS 21847-63-2
Analyt- eller komponentnavne Bephenium Embonate
Forsendelsestilstand Room Temperature
Molekylær formel C23 H14 O6 . 2 C17 H22 N O
Certificeringer/compliance ISO 9001
Synonym Bephenium Embonate, Ammonium, benzyldimethyl(2-phenoxyethyl)-, salt with 4,4'-methylenebis[3-hydroxy-2-naphthoic acid] (2:1) (8CI), Bis[benzyldimethyl(2-phenoxyethyl)ammonium] 4,4'-methylenebis[3-hydroxy-2-naphthoate] (6CI,7CI), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, ion(2-), bis[benzyldimethyl(2-phenoxyethyl)ammonium] (8CI)
InChI formel InChI=1S/C23H16O6.2C17H22NO/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;2*1-18(2,15-16-9-5-3-6-10-16)13-14-19-17-11-7-4-8-12-17/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2*3-12H,13-15H2,1-2H3/q;2*+1/p-2
Løsningstype Neat
SMIL C[N+](C)(CCOc1ccccc1)Cc2ccccc2.C[N+](C)(CCOc3ccccc3)Cc4ccccc4.Oc5c(Cc6c(O)c(cc7ccccc67)C(=O)[O-])c8ccccc8cc5C(=O)[O-]
IUPAC navn benzyl-dimethyl-(2-phenoxyethyl)azanium;4-[(3-carboxylato-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylate
Industry Type Clinical Testing
Molekylvægt (g/mol) 899.08
Mængde 10 mg
Formel vægt 898.4193 g/mol