Learn More
Benzyltriphenylphosphonium chloride, 99% Product Code.: 11407926
Benzyltriphenylphosphonium chloride, 99%
Product Code. 11407926
500 g, 500g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 11407926
Brand: Thermo Scientific Alfa Aesar A14034.36
Request Bulk or Custom Format
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.
Solubility
Soluble in water and alcohol. Insoluble in acetone.
Notes
Hygroscopic. Store under inert gas. Store away from strong oxidizing agents, water/ moisture..
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1100-88-5
Molecular Formula C25H22ClP
Molecular Weight (g/mol) 388.87
MDL Number MFCD00011913
InChI Key USFRYJRPHFMVBZ-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
PubChem CID 70671
IUPAC Name benzyl(triphenyl)phosphanium;chloride
SMILES [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point ∼330°C (decomposition)
Flash Point >300°C (572°F)
Odor Odorless
Quantity 500 g
UN Number UN3464
Beilstein 3599868
Sensitivity Hygroscopic
Solubility Information Soluble in water and alcohol. Insoluble in acetone.
Formula Weight 388.88
Percent Purity 99%
Chemical Name or Material Benzyltriphenylphosphonium chloride
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10249095 Certificate TypeCertificate of Analysis Date25/12/2023 Product CodeA14034.14,A14034.36,A14034.22
Lot Number10226764 Certificate TypeCertificate of Analysis Date17/10/2023 Product CodeA14034.14,A14034.36,A14034.22
Lot Number10244662 Certificate TypeCertificate of Analysis Date17/11/2022 Product CodeA14034.14,A14034.36,A14034.22
Lot NumberC1738B Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA14034.14,A14034.36,A14034.22
Lot Number10226764 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA14034.14,A14034.36,A14034.22
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Product Title
Benzyltriphenylphosphonium chloride, 99% >