Learn More
Benzyltrimethylammonium Chloride, Sigma-Aldrich™
Product Code. 15801591
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity Unit Size
15801591 250 g 250g
1 options
Product Code. 15801591 Supplier Sigma Aldrich Supplier No. 8219610250
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56-93-9
Grade Research
Molecular Formula C6H5CH2(CH3)3NCl
Molecular Weight (g/mol) 185.69
Specifications
Melting Point 243°C
Density 1.07 g/cm3 at 20°C
MDL Number MFCD00011782
Quantity 250 g
Synonym Benzyltrimethylammonium chloride, Trimethylbenzylammonium chloride, Benzyltrimethylammonium chloride