missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-66141
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
56-93-9
C10H16ClN
185.70
MFCD00011782
KXHPPCXNWTUNSB-UHFFFAOYSA-M
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
5963
benzyl(trimethyl)azanium;chloride
[Cl-].C[N+](C)(C)CC1=CC=CC=C1
This item is not returnable. View return policy
56-93-9
C10H16ClN
185.70
MFCD00011782
KXHPPCXNWTUNSB-UHFFFAOYSA-M
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
5963
benzyl(trimethyl)azanium;chloride
[Cl-].C[N+](C)(C)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 239.0°C
Color White to Ivory
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5CH2N(CH3)3Cl
Quantity 50 g
Beilstein 12, 1020
Fieser 01,53
Solubility Information Solubility in water: 800g/L. Other solubilities: soluble in ethanol and butanol,slightly soluble in butyl phthalate and,tributyl phosphate
Viscosity 10 mPa.s (20°C)
Formula Weight 185.7
Percent Purity 98+%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Benzyltrimethylammonium chloride, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'