missing translation for 'onlineSavingsMsg'
Learn More

Benzylpenicillin Procaine, LoGiCal

Product Code. 30461755
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30461755 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461755 Supplier LoGiCal Supplier No. LGCFOR0302.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Procaine benzylpenicillin hydrate
CAS 6130-64-9
Analyt- eller komponentnavne Procaine benzylpenicillin hydrate
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C16 H18 N2 O4 S . C13 H20 N2 O2 . H2 O
Certificeringer/compliance ISO 9001
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- (2S,5R,6R)-, compd. with 2-(diethylamino)ethyl 4-aminobenzoate, hydrate (1:1:1), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-, compd. with 2-(diethylamino)ethyl p-aminobenzoate (1:1), monohydrate (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- (2S,5R,6R)-, compd. with 2-(diethylamino)ethyl 4-aminobenzoate (1:1), monohydrate (9CI), Benzoic acid, 4-amino-, 2-(diethylamino)ethyl ester, mono[(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate], monohydrate (9CI), Benzoic acid, p-amino-, 2-(diethylamino)ethyl ester, mono[3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate], monohydrate (8CI), Benzylpenicillin procaine monohydrate, Procaine penicillin G hydrate, Procaine penicillin G monohydrate, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- [2S-(2alpha,5alpha,6beta)]-, compd. with 2-(diethylamino)ethyl 4-aminobenzoate (1:1), monohydrate, Benzoic acid, 4-amino-, 2-(diethylamino)ethyl ester, mono[[2S-(2alpha,5alpha,6beta)]-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate], monohydrate
InChI formel InChI=1S/C16H18N2O4S.C13H20N2O2.H2O/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);5-8H,3-4,9-10,14H2,1-2H3;1H2/t11-,12+,14-;;/m1../s1
Løsningstype Neat
SMIL O.CCN(CC)CCOC(=O)c1ccc(N)cc1.CC2(C)S[C@@H]3[C@H](NC(=O)Cc4ccccc4)C(=O)N3[C@H]2C(=O)O
IUPAC navn 2-(diethylamino)ethyl 4-aminobenzoate;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;hydrate
Industry Type Clinical Testing
Molekylvægt (g/mol) 588.72
Mængde 10 mg
Formel vægt 588.2618 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.