Få mere at vide
Benzylpenicillin Potassium EP Impurity C (as Sodium Salt), Mikromol™
Beskrivelse
Benzylpenicillin Potassium EP Impurity C (as Sodium Salt)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Hydroxybenzylpenicillin Sodium Salt |
| CAS | 5985-13-7 |
| Analyt- eller komponentnavne | 4-Hydroxybenzylpenicillin Sodium Salt |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C16 H17 N2 O5 S . Na |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzylpenicillin Potassium Imp. C (EP), Benzylpenicillin Imp. C (EP), Benzylpenicillin Potassium Imp. C (EP) as Sodium Salt, (2S,5R,6R)-6-[[(4-Hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Sodium Salt, 4-Hydroxybenzylpenicillin Sodium Salt, Benzylpenicillin Benzathine Impurity G as Sodium Salt, Benzylpenicillin Potassium Impurity C as Sodium Salt, Benzylpenicillin Procaine Impurity F as Sodium Salt, Benzylpenicillin Impurity C as Sodium Salt |
| InChI formel | InChI=1S/C16H18N2O5S.Na/c1-16(2)12(15(22)23)18-13(21)11(14(18)24-16)17-10(20)7-8-3-5-9(19)6-4-8;/h3-6,11-12,14,19H,7H2,1-2H3,(H,17,20)(H,22,23);/q;+1/p-1/t11-,12+,14-;/m1./s1 |
| SMIL | [Na+].CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccc(O)cc3)C(=O)N2[C@H]1C(=O)[O-] |
| IUPAC navn | sodium;(2S,5R,6R)-6-[[2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| Vis mere |