Få mere at vide
Benzylpenicillin 1000 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Benzylpenicillin |
| CAS | 61-33-6 |
| Analyt- eller komponentnavne | Benzylpenicillin |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C16 H18 N2 O4 S |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]- (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-(2-phenylacetamido)- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- [2S-(2α,5α,6β)]-, Penicillin G (6CI), (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (2S,5R,6R)-3,3-Dimethyl-6-(2-(phenylacetylamino)-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (5R,6R)-Benzylpenicillin, Allpen, Benzyl-6-aminopenicillinic acid, Benzylpenicillin, Benzylpenicillin G, Benzylpenicillinic acid, Cilloral, Cilopen, Crysticillin 300 AS, Dropcillin, Free benzylpenicillin, Free penicillin G, Free penicillin II, Gelacillin, Liquacillin, NSC 193396, Penicillin, Penicillin, (phenylmethyl)-, Penicillinic acid, (phenylmethyl)-, Pharmacillin, Phenylacetamidopenicillanic acid, Phenylacetyl-6-aminopenicillanic acid, Pradupen, Specilline G, Ursopen |
| InChI formel | InChI=1S/C16H18N2O4S/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22)/t11-,12+,14-/m1/s1 |
| SMIL | CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(=O)O |
| Vis mere |