Få mere at vide
Benzylidene Camphor Sulfonic Acid, TRC
Artikelnummer. 30012091
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012091 50 mg
30008079 250 mg
30007642 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012091 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28003850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56039-58-8
Molekylær formel C17 H20 O4 S
Molekylvægt (g/mol) 320.4
Synonym 4-[(4,7,7-Trimethyl-3-oxobicyclo[2.2.1]hept-2-ylidene)methyl]benzenesulfonic acid, 4-(2-Oxo-3-bornylidenemethyl)benzenesulfonic acid, 4-(2-Oxo-3-bornylidennemethyl)benzosulfonic Acid, 4'-Sulfo-3-benzylidenecamphor, Mexoryl SL, p-[(2-Oxo-3-bornylidene)methyl]benzenesulfonic acid, Benzenesulfonic acid, 4-[(4,7,7-trimethyl-3-oxobicyclo[2.2.1]hept-2-ylidene)methyl]-
IUPAC navn 4-[(Z)-[(1S,4R)-4,7,7-trimethyl-3-oxo-2-bicyclo[2.2.1]heptanylidene]methyl]benzenesulfonic acid
SMIL CC1(C)[C@@H]2CC[C@@]1(C)C(=O)/C/2=C\c3ccc(cc3)S(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Benzylidene Camphor Sulfonic Acid
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C17H20O4S/c1-16(2)14-8-9-17(16,3)15(18)13(14)10-11-4-6-12(7-5-11)22(19,20)21/h4-7,10,14H,8-9H2,1-3H3,(H,19,20,21)/b13-10-/t14-,17+/m1/s1
Formel vægt 320.1082
Procent renhed >95%