missing translation for 'onlineSavingsMsg'
Få mere at vide

Benzyldodecyldimethylammonium Bromide, TRC

Artikelnummer. 30346312
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30346312 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30346312 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2797801G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Benzyldodecyldimethylammonium Bromide
CAS 7281-04-1
Renhedsgrad noter HPLC
Molekylær formel C21 H38 N . Br
Mængde 1 g
Synonym Benzalkonium Bromide (C12), Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, bromide (1:1), Ammonium, benzyldodecyldimethyl-, bromide (8CI), Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, bromide (9CI), Benzyldodecyldimethylammonium bromide (6CI,7CI), AACL 6, Amonyl BR 1244, Bacfor BL, Benzododecinium bromide, Benzyldimethyldodecylammonium bromide, Benzyldimethyllaurylammonium bromide, Benzyllauryldimethylammonium bromide, Cequartyl 50, Dimethylbenzyldodecylammonium bromide, Dimethylbenzyllaurylammonium bromide, Dimethyldodecylbenzylammonium bromide, Dimethyllaurylbenzylammonium bromide, Dodecylbenzyldimethylammonium bromide, Dodecyldimethylbenzylammonium bromide, Lauralkonium bromide, Lauryldimethylbenzylammonium bromide, N,N-Dimethyl-N-benzyldodecylammonium bromide, N,N-Dimethyl-N-dodecylbenzylammonium bromide, N-Benzyl-N,N-dimethyl-N-dodecylammonium bromide, Sinnoquat BL 80, Sinnoquat BL 95, Sterinol, Xinjieermie
InChI formel InChI=1S/C21H38N.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1
SMIL [Br-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
IUPAC navn benzyl-dodecyl-dimethylazanium;bromide
Molekylvægt (g/mol) 384.44
Formel vægt 383.2188
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.