missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Benzyldodecyldimethylammonium Bromide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Benzyldodecyldimethylammonium Bromide |
| CAS | 7281-04-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C21 H38 N . Br |
| Mængde | 100 mg |
| Synonym | Benzalkonium Bromide (C12), Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, bromide (1:1), Ammonium, benzyldodecyldimethyl-, bromide (8CI), Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, bromide (9CI), Benzyldodecyldimethylammonium bromide (6CI,7CI), AACL 6, Amonyl BR 1244, Bacfor BL, Benzododecinium bromide, Benzyldimethyldodecylammonium bromide, Benzyldimethyllaurylammonium bromide, Benzyllauryldimethylammonium bromide, Cequartyl 50, Dimethylbenzyldodecylammonium bromide, Dimethylbenzyllaurylammonium bromide, Dimethyldodecylbenzylammonium bromide, Dimethyllaurylbenzylammonium bromide, Dodecylbenzyldimethylammonium bromide, Dodecyldimethylbenzylammonium bromide, Lauralkonium bromide, Lauryldimethylbenzylammonium bromide, N,N-Dimethyl-N-benzyldodecylammonium bromide, N,N-Dimethyl-N-dodecylbenzylammonium bromide, N-Benzyl-N,N-dimethyl-N-dodecylammonium bromide, Sinnoquat BL 80, Sinnoquat BL 95, Sterinol, Xinjieermie |
| InChI formel | InChI=1S/C21H38N.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1 |
| SMIL | [Br-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1 |
| IUPAC navn | benzyl-dodecyl-dimethylazanium;bromide |
| Molekylvægt (g/mol) | 384.44 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?