missing translation for 'onlineSavingsMsg'
Få mere at vide

Benzyldimethylsilane, 96%, BLD Pharm™

Artikelnummer. 30779794
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
5 g
25 g
Pakningsstørrelse:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde Unit Size
30779794 5 g 5g
30769238 1 g 1g
30775642 25 g 25g
30779446 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30779794 Supplier BLD Pharm Supplier No. BD1162835g
 Begrænset vare
Forventet forsendelse: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale Benzyldimethylsilane
CAS 1631-70-5
Molekylær formel C9H14Si
MDL nummer MFCD00039792
Mængde 5 g
SMIL C[SiH](CC1=CC=CC=C1)C
Molekylvægt (g/mol) 150.29
Procent renhed 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.