Få mere at vide
Benzylamine-d7, TRC
Artikelnummer. 30339313
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30339313 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339313 Leverandør Toronto Research Chemicals Leverandørnr. TRCB224863100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzylamine-d7
CAS 167750-79-0
Molekylær formel C7 D7 H2 N
Mængde 100 mg
Synonym Benzene-d5-methan-d2-amine (9CI), Benzyl-d7-amine, Benzylamine-d7, Benzylamine-D7
InChI formel InChI=1S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5H,6,8H2/i1D,2D,3D,4D,5D,6D2
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])N
IUPAC navn dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanamine
Molekylvægt (g/mol) 114.196
Formel vægt 114.117