missing translation for 'onlineSavingsMsg'
Få mere at vide

Benzylamine-d7, TRC

Artikelnummer. 30343756
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343756 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343756 Leverandør Toronto Research Chemicals Leverandørnr. TRCB22486310MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Benzylamine-d7
CAS 167750-79-0
Molekylær formel C7 D7 H2 N
Mængde 10 mg
Synonym Benzene-d5-methan-d2-amine (9CI), Benzyl-d7-amine, Benzylamine-d7, Benzylamine-D7
InChI formel InChI=1S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5H,6,8H2/i1D,2D,3D,4D,5D,6D2
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])N
IUPAC navn dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanamine
Molekylvægt (g/mol) 114.196
Formel vægt 114.117
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.