missing translation for 'onlineSavingsMsg'
Learn More

Benzyl N-Diphenylmethyl-2-azetidinecarboxylate, TRC

Product Code. 30344237
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30344237 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344237 Supplier Toronto Research Chemicals Supplier No. TRCB2763151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Benzyl N-Diphenylmethyl-2-azetidinecarboxylate
CAS 22742-42-3
Molekylær formel C24 H23 N O2
Mængde 1 g
InChI formel InChI=1S/C24H23NO2/c26-24(27-18-19-10-4-1-5-11-19)22-16-17-25(22)23(20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-15,22-23H,16-18H2
SMIL O=C(OCc1ccccc1)C2CCN2C(c3ccccc3)c4ccccc4
IUPAC navn benzyl 1-benzhydrylazetidine-2-carboxylate
Molekylvægt (g/mol) 357.44
Formel vægt 357.1729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.