Learn More
Benzyl methyl ether, 97%
Product Code. 11405497
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405497 10 g 10g
11415497 50 g 50g
11425497 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11405497 Supplier Thermo Scientific Alfa Aesar Supplier No. B23490.09
 Restricted Item
På lager
Add to basket
CAS: 538-86-3 | C8H10O | 122.167 g/mol
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 538-86-3
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00025901
InChI Key GQKZBCPTCWJTAS-UHFFFAOYSA-N
Synonym benzyl methyl ether, methoxymethyl benzene, methyl benzyl ether, benzene, methoxymethyl, ether, benzyl methyl, benzylmethylether, unii-f22rls78bd, .alpha.-methoxytoluene, f22rls78bd, methoxyphenylmethane
PubChem CID 10869
IUPAC Name methoxymethylbenzene
SMILES COCC1=CC=CC=C1
Specifications
Melting Point -53°C
Density 0.94
Boiling Point 173°C to 175°C
Flash Point 56°C (133°F)
Refractive Index 1.5015
Quantity 10 g
UN Number UN3271
Merck Index 14,1140
Solubility Information Insoluble in water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material Benzyl methyl ether
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl methyl ether
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Benzyl methyl ether