missing translation for 'onlineSavingsMsg'
Learn More

Benzyl L-lactate, 97%, Thermo Scientific Chemicals

Catalog Number p-7058424
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
56777-24-3
C10H12O3
180.203
MFCD00209546
ZYTLPUIDJRKAAM-QMMMGPOBSA-N
benzyl s---lactate, benzyl l-lactate, benzyl s-lactate, s---lactic acid benzyl ester, benzyl 2s-2-hydroxypropanoate, benzyl-l-lactate, s-benzyllactate, s-benzyl lactate, s-lactic acid benzyl ester, 2s-lactic acid benzyl ester
11052238
benzyl (2S)-2-hydroxypropanoate
CC(C(=O)OCC1=CC=CC=C1)O
This item is not returnable. View return policy
56777-24-3
C10H12O3
180.203
MFCD00209546
ZYTLPUIDJRKAAM-QMMMGPOBSA-N
benzyl s---lactate, benzyl l-lactate, benzyl s-lactate, s---lactic acid benzyl ester, benzyl 2s-2-hydroxypropanoate, benzyl-l-lactate, s-benzyllactate, s-benzyl lactate, s-lactic acid benzyl ester, 2s-lactic acid benzyl ester
11052238
benzyl (2S)-2-hydroxypropanoate
CC(C(=O)OCC1=CC=CC=C1)O

Benzyl L-lactate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl L-lactate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.12
Boiling Point 105°C to 109°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.513
Quantity 5 g
Beilstein 2048925
Solubility Information Not miscible or difficult to mix in water.
Optical Rotation −12.9° (c=3.1 in Methanol)
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Benzyl L-lactate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'