missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isovalerate, 98%, Thermo Scientific™

Catalog Number p-7027820
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
103-38-8
C12H16O2
192.26
MFCD00048348
HVJKZICIMIWFCP-UHFFFAOYSA-N
benzyl isovalerate, benzyl 3-methylbutyrate, benzyl isopentanoate, butanoic acid, 3-methyl-, phenylmethyl ester, isovaleric acid, benzyl ester, phenylmethyl 3-methylbutanoate, benzyl 3-methyl butyrate, benzyl isovalerianate, isopropyl acetic acid, benzyl ester, isopentanoic acid, phenylmethyl ester
7651
benzyl 3-methylbutanoate
CC(C)CC(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
103-38-8
C12H16O2
192.26
MFCD00048348
HVJKZICIMIWFCP-UHFFFAOYSA-N
benzyl isovalerate, benzyl 3-methylbutyrate, benzyl isopentanoate, butanoic acid, 3-methyl-, phenylmethyl ester, isovaleric acid, benzyl ester, phenylmethyl 3-methylbutanoate, benzyl 3-methyl butyrate, benzyl isovalerianate, isopropyl acetic acid, benzyl ester, isopentanoic acid, phenylmethyl ester
7651
benzyl 3-methylbutanoate
CC(C)CC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.985
Boiling Point 245°C
Quantity 50 g
Formula Weight 192.26
Percent Purity 98%
Chemical Name or Material Benzyl isovalerate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'