Få mere at vide
Benzyl Isothiocyanate-d7, TRC
Artikelnummer. 30240649
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240649 5 mg
30240513 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240649 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2815025MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246818-63-2
Molekylær formel C8D7NS
Molekylvægt (g/mol) 156.26
Synonym Isothiocyanic Acid (Benzyl-d7) Ester, (Isothiocyanatomethyl)benzene-d7, 1-(Isothiocyanatomethyl)benzene-d7, AB 2-d7, (Benzyl-d7) Mustard Oil, NSC 118976-d7, (Phenylmethyl-d7) Isothiocyanate, Tromacaps-d7, Tromalyt-d7, Urogran-d7;
IUPAC navn 1,2,3,4,5-pentadeuterio-6-[dideuterio(isothiocyanato)methyl]benzene
SMIL [2H]C1=C(C([2H])([2H])N=C=S)C([2H])=C([2H])C([2H])=C1[2H]
Tekniske data
Kemisk navn eller materiale Benzyl Isothiocyanate-d7
Mængde 5 mg
InChI formel InChI=1S/C8H7NS/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2/i1D,2D,3D,4D,5D,6D2