Få mere at vide
Benzyl-d7 Chloride, CDN
Artikelnummer. 30328452
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30328452 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328452 Leverandør CDN Leverandørnr. CDND03301G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-d7 Chloride
CAS 59502-05-5
Molekylær formel C7 D7 Cl
Mængde 1 g
InChI formel InChI=1 S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5 H,6H2/i1D,2 D,3 D,4 D,5 D,6D2
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])Cl
IUPAC navn 1-[chloro(dideuterio)methyl]-2,3,4,5,6-pentadeuterio-benzene
Molekylvægt (g/mol) 133.63
Formel vægt 133.0676 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity