Få mere at vide
Benzyl-d7 Bromide, TRC
Artikelnummer. 30343119
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30343119 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343119 Leverandør Toronto Research Chemicals Leverandørnr. TRCB233653100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-d7 Bromide
CAS 35656-93-0
Molekylær formel C7 D7 Br
Mængde 100 mg
Synonym Benzene-1,2,3,4,5-d5, 6-(bromomethyl-d2)- (ACI), 6-(Bromomethyl-d2)benzene-1,2,3,4,5-d5 (ACI), Benzene-d5, (bromomethyl-d2)- (9CI), (Bromomethyl-d2)benzene-d5, 1-[Bromo(dideuterio)methyl]-2,3,4,5,6-pentadeuteriobenzene, Benzyl-d7 bromide, Benzyl bromide-d7, Benzyl bromide-D7
InChI formel InChI=1S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D,6D2
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])Br
IUPAC navn 1-[bromo(dideuterio)methyl]-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 178.0776
Formel vægt 177.0171