Få mere at vide
Benzyl-d7 Bromide, CDN
Artikelnummer. 30313099
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313099 0.5 g
30321458 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313099 Leverandør CDN Leverandørnr. CDND53650.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-d7 Bromide
CAS 35656-93-0
Molekylær formel C7 D7 Br
Mængde 0.5 g
Synonym Benzene-1,2,3,4,5-d5, 6-(bromomethyl-d2)- (ACI), 6-(Bromomethyl-d2)benzene-1,2,3,4,5-d5 (ACI), Benzene-d5, (bromomethyl-d2)- (9 CI), (Bromomethyl-d2)benzene-d5, 1-[Bromo(dideuterio)methyl]-2,3,4,5,6-pentadeuteriobenzene, Benzyl-d7 bromide, Benzyl bromide-d7, Benzyl bromide-D7
InChI formel InChI=1 S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5 H,6H2/i1D,2 D,3 D,4 D,5 D,6D2
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])Br
IUPAC navn 1-[bromo(dideuterio)methyl]-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 178.0776
Formel vægt 177.0171 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity