Få mere at vide
Benzyl-d7-amine HCl, CDN
Artikelnummer. 30326937
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326937 0.25 g
30320005 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326937 Leverandør CDN Leverandørnr. CDND52260.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzylamine-D7 Hydrochloride
CAS 352431-27-7
Molekylær formel C7 D7 H2 N . Cl H
Mængde 0.25 g
Synonym Benzene-d5-methan-d2-amine, hydrochloride (9 CI), Benzene-d5-methan-d2-amine hydrochloride, Benzyl-d7-amine hydrochloride, Benzylamine-d7 hydrochloride, Benzylamine-D7 hydrochloride
InChI formel InChI=1 S/C7H9N.ClH/c8-6-7-4-2-1-3-5-7;/h1-5 H,6,8H2;1 H/i1D,2 D,3 D,4 D,5 D,6D2;
SMIL Cl.[2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])N
IUPAC navn dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanamine;hydrochloride
Molekylvægt (g/mol) 150.6571
Formel vægt 150.0941 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity