Få mere at vide
Benzyl-d5 Chloride, TRC
Artikelnummer. 30347361
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30347361 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347361 Leverandør Toronto Research Chemicals Leverandørnr. TRCB23367725MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-d5 Chloride
CAS 68661-11-0
Molekylær formel C7 D5 H2 Cl
Mængde 25 mg
Synonym 6-(Chloromethyl)benzene-1,2,3,4,5-d5, alpha-Chlorotoluene-d5, 1-Chloromethylbenzene-d5, Benzyl Chloride-d5, Chlorophenylmethane-d5, NSC 8043-d5, Phenylmethyl Chloride-d5, Tolyl Chloride-d5, ω-Chlorotoluene-d5, Benzene-1,2,3,4,5-d5, 6-(chloromethyl)-, Benzene-d5, (chloromethyl)- (9CI), Benzyl chloride (ring-D5)
InChI formel InChI=1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(CCl)c([2H])c1[2H]
IUPAC navn 1-(chloromethyl)-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 131.61
Formel vægt 131.055