Få mere at vide
Benzyl-d5 Bromide, TRC
Artikelnummer. 30340338
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30340338 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340338 Leverandør Toronto Research Chemicals Leverandørnr. TRCB233652250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-d5 Bromide
CAS 71258-22-5
Molekylær formel C7 2H5 H2 Br
Mængde 250 mg
Synonym Benzene-1,2,3,4,5-d5, 6-(bromomethyl)-, Benzene-d5, (bromomethyl)- (9CI), 6-(Bromomethyl)benzene-1,2,3,4,5-d5, 1-(Bromomethyl)-2,3,4,5,6-pentadeuteriobenzene, 2,3,4,5,6-Pentadeuteriobenzyl bromide, Benzyl Bromide-D5 (phenyl-D5), Benzyl-d5 Bromide, Benzyl-2,3,4,5,6-d5 Bromide
InChI formel InChI=1S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(CBr)c([2H])c1[2H]
IUPAC navn 1-(bromomethyl)-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 176.07
Formel vægt 175.0045