Learn More
Benzyl Chloride, Sigma-Aldrich™
Product Code. 15874051
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874051 2.5 L 2.5 L
15854051 100 mL 100mL
15864051 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15874051 Supplier Sigma Aldrich Supplier No. 8018092500
 Restricted Item
Forventet forsendelse: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-44-7
Grade Research
Molecular Weight (g/mol) 126.58
Specifications
Melting Point -39°C
Density 1.10 g/cm3 at 20°C
Boiling Point 179.3°C/1013 hPa
MDL Number MFCD00000889
Quantity 2.5 L
Synonym Benzyl chloride, Benzyl chloride