Få mere at vide
Benzyl Beta-D-Lactoside, TRC
Artikelnummer. 30332875
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30332875 250 mg
30330953 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30332875 Leverandør Toronto Research Chemicals Leverandørnr. TRCB281625250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl beta-D-lactoside
CAS 18404-72-3
Molekylær formel C19 H28 O11
Mængde 250 mg
InChI formel InChI=1S/C19H28O11/c20-6-10-12(22)13(23)15(25)19(28-10)30-17-11(7-21)29-18(16(26)14(17)24)27-8-9-4-2-1-3-5-9/h1-5,10-26H,6-8H2/t10-,11-,12+,13+,14-,15-,16-,17-,18-,19+/m1/s1
SMIL OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OCc3ccccc3)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
IUPAC navn (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-phenylmethoxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvægt (g/mol) 432.42
Formel vægt 432.1632