Få mere at vide
Benzyl Benzoate-d12, TRC
Artikelnummer. 30348542
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30348542 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348542 Leverandør Toronto Research Chemicals Leverandørnr. TRCB23095225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl Benzoate-d12
CAS 352431-26-6
Molekylær formel C14 D12 O2
Mængde 25 mg
Synonym Benzoic-d5 acid, phenyl-d5-methyl-d2 ester (9CI), Benzyl Benzoate-d12, Benzyl Benzoate-D12, Benzoic-d5 acid phenyl-d5-methyl-d2 ester, Benzoic-d5 acid benzyl-d7 ester
InChI formel InChI=1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D2
SMIL [2H]c1c([2H])c([2H])c(C(=O)OC([2H])([2H])c2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]
IUPAC navn [dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl] 2,3,4,5,6-pentadeuteriobenzoate
Molekylvægt (g/mol) 224.3178
Formel vægt 224.1591