Få mere at vide
Benzyl-alpha,alpha-d2-amine, CDN
Artikelnummer. 30327953
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327953 0.5 g
30324252 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327953 Leverandør CDN Leverandørnr. CDND61330.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzylamine-alpha,alpha-D2
CAS 15185-02-1
Molekylær formel C7 D2 H7 N
Mængde 0.5 g
Synonym Benzenemethan-d2-amine (9 CI), Benzyl-α,α-d2-amine (8 CI), α,α-Dideuteriobenzylamine, α-Dideuteriobenzylamine, Benzylamine-d2, Benzylamine-α,α-d2, Benzylamine-α,α-D2
InChI formel InChI=1 S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5 H,6,8H2/i6D2
SMIL [2 H]C([2 H])(N)c1ccccc1
IUPAC navn dideuterio(phenyl)methanamine
Molekylvægt (g/mol) 109.1654
Formel vægt 109.0861 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity