Få mere at vide
Benzyl-Alpha,Alpha-d2 Alcohol, TRC
Artikelnummer. 30451719
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30451719 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451719 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53663610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-Alpha,Alpha-d2 Alcohol
CAS 21175-64-4
Molekylær formel C7H6D2O
Mængde 10 mg
Synonym Benzyl-α,α-d2 alcohol (6CI','7CI','8CI), 1,1-d2-Phenylmethanol, [α,α-2H2]Benzyl alcohol, Benzenemethanol-α,α-d2, α,α-Dideuteriobenzyl alcohol
InChI formel InChI=1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2/i6D2
SMIL OC([2H])([2H])C1=CC=CC=C1
IUPAC navn phenylmethan-d2-ol
Molekylvægt (g/mol) 110.15
Formel vægt 110.07