Få mere at vide
Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate, Mikromol™
Artikelnummer. 30463512
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463512 4 x 25 mg
30050664 25 mg
17928714 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30463512 Leverandør Mikromol Leverandørnr. MM3300.1800254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate
CAS 168828-81-7
Analyt- eller komponentnavne Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate
Forsendelsestilstand Room Temperature
Molekylær formel C18 H19 F N2 O3
Certificeringer/compliance ISO 17025
Synonym N-[3-Fluoro-4-(4-morpholinyl)phenyl]carbamic Acid Phenylmethyl Ester, Carbamic acid, [3-fluoro-4-(4-morpholinyl)phenyl]-, phenylmethyl ester (9CI), (3-Fluoro-4-morpholin-4-ylphenyl)carbamic acid benzyl ester, Benzyl 3-fluoro-4-(4-morpholinyl)phenylcarbamate, N-(Benzyloxycarbonyl)-3-fluoro-4-(4-morpholinyl)aniline, N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline, [3-Fluoro-4-(4-morpholinyl)phenyl]carbamic acid phenylmethyl ester
InChI formel InChI=1S/C18H19FN2O3/c19-16-12-15(6-7-17(16)21-8-10-23-11-9-21)20-18(22)24-13-14-4-2-1-3-5-14/h1-7,12H,8-11,13H2,(H,20,22)
Løsningstype Neat
SMIL Fc1cc(NC(=O)OCc2ccccc2)ccc1N3CCOCC3
IUPAC navn benzyl N-(3-fluoro-4-morpholin-4-ylphenyl)carbamate
Molekylvægt (g/mol) 330.35
Mængde 4 x 25 mg
Formel vægt 330.138 g/mol