missing translation for 'onlineSavingsMsg'
Få mere at vide

Benzyl 2,3,4-Tri-O-benzyl-D-glucuronate, TRC

Artikelnummer. 30344166
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30344166 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30344166 Leverandør Toronto Research Chemicals Leverandørnr. TRCB315885250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Benzyl 2,3,4-Tri-O-benzyl-D-glucuronate
CAS 53684-90-5
Molekylær formel C34 H34 O7
Mængde 250 mg
InChI formel InChI=1S/C34H34O7/c35-33(40-24-28-19-11-4-12-20-28)32-30(38-22-26-15-7-2-8-16-26)29(37-21-25-13-5-1-6-14-25)31(34(36)41-32)39-23-27-17-9-3-10-18-27/h1-20,29-32,34,36H,21-24H2/t29-,30-,31+,32-,34?/m0/s1
SMIL OC1O[C@@H]([C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@H]1OCc4ccccc4)C(=O)OCc5ccccc5
IUPAC navn benzyl (2S,3S,4S,5R)-6-hydroxy-3,4,5-tris(phenylmethoxy)oxane-2-carboxylate
Molekylvægt (g/mol) 554.63
Formel vægt 554.2305
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.