Få mere at vide
Benzyl-2,3,4,5,6-d5-dimethyl-n-tetradecylammonium Bromide, TRC
Artikelnummer. 30345001
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30345001 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345001 Leverandør Toronto Research Chemicals Leverandørnr. TRCB27630225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-2,3,4,5,6-d5-dimethyl-n-tetradecylammonium Bromide
CAS 1515861-68-3
Molekylær formel C23 D5 H37 N . Br
Mængde 25 mg
Synonym Benzene-2,3,4,5,6-d5-methanaminium, N,N-dimethyl-N-tetradecyl-, bromide (1:1) (ACI), N-Tetradecyl-N,N-dimethylbenzene-d5-methanaminium bromide, (Benzyl-2,3,4,5-d5)dimethyltetradecylammonium bromide, Benzyldimethyltetradecylammonium-D5 (phenyl-D5) Bromide
InChI formel InChI=1S/C23H42N.BrH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1/i15D,16D,17D,19D,20D;
SMIL [Br-].[2H]c1c([2H])c([2H])c(C[N+](C)(C)CCCCCCCCCCCCCC)c([2H])c1[2H]
IUPAC navn dimethyl-[(2,3,4,5,6-pentadeuteriophenyl)methyl]-tetradecylazanium;bromide
Molekylvægt (g/mol) 417.5211
Formel vægt 416.2814