Få mere at vide
Benzyl-2,3,4,5,6-d5 Chloride, CDN
Artikelnummer. 30308806
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30308806 0.5 g
30311392 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308806 Leverandør CDN Leverandørnr. CDND16520.5G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl-d5 Chloride
CAS 68661-11-0
Molekylær formel C7 D5 H2 Cl
Mængde 0.5 g
Synonym 6-(Chloromethyl)benzene-1,2,3,4,5-d5, alpha-Chlorotoluene-d5, 1-Chloromethylbenzene-d5, Benzyl Chloride-d5, Chlorophenylmethane-d5, Benzene-d5, (chloromethyl)- (9 CI), Benzene-d5, (chloromethyl)- (9 CI), Benzene-d5, (chloromethyl)- (9 CI), Benzene-d5, (chloromethyl)- (9 CI), Benzene-d5, (chloromethyl)- (9 CI), Benzene-d5, (chloromethyl)- (9 CI), Benzene-d5, (chloromethyl)- (9 CI)
InChI formel InChI=1 S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5 H,6H2/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(CCl)c([2 H])c1[2 H]
IUPAC navn 1-(chloromethyl)-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 131.61
Formel vægt 131.055 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity