Få mere at vide
Benzyl [1-[4-[[(4-Fluorobenzyl)amino]carbonyl]-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]-1-methylethyl]carbamate-d3, TRC
Artikelnummer. 30338067
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30338067 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338067 Leverandør Toronto Research Chemicals Leverandørnr. TRCB27659210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzyl (1-(4-(((4-Fluorobenzyl)amino)carbonyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl)-1-methylethyl)carbamate-d3
CAS 1189916-86-6
Molekylær formel C24H22D3FN4O5
Mængde 10 mg
Synonym N-[1-[4-[[[(4-Fluorophenyl)methyl]amino]carbonyl]-1,6-dihydro-5-hydroxy-1-(methyl-d3)-6-oxo-2-pyrimidinyl]-1-methylethyl]carbamic Acid Phenylmethyl Ester
InChI formel InChI=1S/C24H25FN4O5/c1-24(2,28-23(33)34-14-16-7-5-4-6-8-16)22-27-18(19(30)21(32)29(22)3)20(31)26-13-15-9-11-17(25)12-10-15/h4-12,30H,13-14H2,1-3H3,(H,26,31)(H,28,33)/i3D3
SMIL [2H]C([2H])([2H])N1C(=O)C(=C(N=C1C(C)(C)NC(=O)OCc2ccccc2)C(=O)NCc3ccc(F)cc3)O
IUPAC navn benzyl N-[2-[4-[(4-fluorophenyl)methylcarbamoyl]-5-hydroxy-6-oxo-1-(trideuteriomethyl)pyrimidin-2-yl]propan-2-yl]carbamate
Molekylvægt (g/mol) 471.5
Formel vægt 471.1997