Få mere at vide
Benzydamine-d6 N-Oxide, TRC
Artikelnummer. 30336678
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30336678 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336678 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2099621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzydamine-d6 N-Oxide
CAS 1246820-03-0
Molekylær formel C19H17D6N3O2
Mængde 1 mg
Synonym N,N-(Dimethyl-d6)-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine
InChI formel InChI=1S/C19H23N3O2/c1-22(2,23)13-8-14-24-19-17-11-6-7-12-18(17)21(20-19)15-16-9-4-3-5-10-16/h3-7,9-12H,8,13-15H2,1-2H3/i1D3,2D3
SMIL O=N(C([2H])([2H])[2H])(C([2H])([2H])[2H])CCCOC1=NN(CC2=CC=CC=C2)C3=CC=CC=C31
IUPAC navn 3-(1-benzylindazol-3-yl)oxy-N,N-bis(trideuteriomethyl)propan-1-amine oxide
Molekylvægt (g/mol) 331.44